Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SUPELCOGEL™ 8H, 8% Crosslinked HPLC Guard Column, 9 μm Particle Size

Catalog No. 111008705 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter: 4.6 mm; Length: 5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-100-8705 Diameter: 4.6 mm; Length: 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-8705 Supplier MilliporeSigma™ Supelco™ Supplier No. 59251U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Offers excellent separation of many common samples.

SUPELCOGEL™ 8 Column Series (manufactured with 8% XL PS-DVB):The majority of columns on the market for the analysis of carbohydrates, organic acids, and alcohols, such as Aminex™, utilize an 8% XL PS-DVB. The 8% polymer offers excellent separation of many common samples. Advantages: 8% polymers combine durability and good resolution. Due to higher cross-linking than 6% XL polymers, columns packed with 8% polymer can be used at higher flow rates therefore decreasing analysis times.Disadvantages: 8% polymers are limited in separating carbohydrates larger than trisaccharides due to lower MW exclusion limits. Compared to the 6% XL polymer, for some types of carbohydrate and organic acid mixtures, the 8% shows less resolution due to selectivity differences. Molecular Weight Exclusion Limit: 1200 minimum. Wet Capacity meq/mL: 1.7 - 1.9. Flow Rate Recommendation: 0.4 - 1.2 ml/min.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 9 μm
Length (Metric) 5 cm
Type Guard Column
Diameter (Metric) Inner 4.6 mm
Crosslinking 0.08
Phase Ion Exclusion
For Use With (Application) HPLC
Product Line SUPELCOGEL

SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.