Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SUPELCOSIL™ Suplex™ pKb-100 Supelguard Cartridge, 5 μm Particle Size

Catalog No. 111003881 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter: 4 mm; Length: 2 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-100-3881 Diameter: 4 mm; Length: 2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3881 Supplier MilliporeSigma™ Supelco™ Supplier No. 59541U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suplex™ pKb-100 columns feature the same bonded phase functionality as SUPELCOSIL™ LC-ABZ columns.

Suplex™ pKb-100 columns feature the same bonded phase functionality as SUPELCOSIL™ LC-ABZ columns. These specially deactivated columns differ in that Suplex™ pKb-100 is not endcapped, while SUPELCOSIL™ LC-ABZ is endcapped. The absence of the end-capping reagent results in better performance from Suplex™ pKb-100 for the strongest basic compounds, while LC-ABZ is preferred when the sample also contains acids and/or zwitterions.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2/Pk.
Particle Size 5 μm
Length (Metric) 2 cm
Pore Size 120 Å
Type Guard Cartridge
Diameter (Metric) Inner 4 mm
Phase Reversed Phase
For Use With (Application) HPLC
Product Line SUPELCOSIL Suplex

SUPELCOSIL and Suplex are trademarks of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.