Learn More
MilliporeSigma™ Supelco™ Supelpure™-O Oxygen/Moisture Trap
Oxygen-removing catalysts that can reduce oxygen to less than 2 ppb when the level in the incoming gas does not exceed 10 ppm
Supplier:  MilliporeSigma™ Supelco™ 503088
 View more versions of this product
Catalog No. 11-100-3761
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
750 cc
Oxygen/Moisture Trap
1/4 in.
41.9 cm
59 mm
Supelpure™
Removing Oxygen and Water Vapor From Incoming Gas
16.5 in.
2 5/16 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Supelpure is a registered trademark of Sigma-Aldrich Co. LLC