Learn More
MilliporeSigma™ Supelco™ Supelpure™-O Oxygen/Moisture Trap
Oxygen-removing catalysts that can reduce oxygen to less than 2 ppb when the level in the incoming gas does not exceed 10 ppm
Supplier: MilliporeSigma™ Supelco™ 22450U
Description
Specifications
120 cc | |
Oxygen/Moisture Trap | |
1/4 in. | |
28.2 cm | |
35 mm |
Supelpure™ | |
Removing Oxygen and Water Vapor From Incoming Gas | |
11.125 in. | |
1 3/8 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Supelpure is a registered trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?Share a Content Correction