Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-1 Packed Column Ferrule, 1/4 in.
Catalog No. 111016347 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-101-6347 10/Pk.
11-100-9359 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6347 Supplier MilliporeSigma™ Supelco™ Supplier No. 22086U
Add to Cart
Add to Cart
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only. Material Composition: Ceramic-filled PTFE. Maximum Temperature: 250°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/4 in.
Diameter (English) Inner 1/4 in.
Packaging Quantity 10/Pk.
Description Ferrule Type: Packed Column
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC