Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-1 Packed Column Ferrule, 1/8 in.
Catalog No. 111009471 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
100/Pk.
Catalog No. Packaging Quantity
11-100-9471 100/Pk.
1 options
Catalog No. 11-100-9471 Supplier MilliporeSigma™ Supelco™ Supplier No. 22309
Add to Cart
Edge
Add to Cart
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only. Material Composition: Ceramic-filled PTFE. Maximum Temperature: 250°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/8 in.
Diameter (English) Inner 1/8 in.
Packaging Quantity 100/Pk.
Description Ferrule Type: Packed Column
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC