Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2 Packed Column Ferrule, 1/4 in.
Catalog No. 111009525 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-100-9525 50/Pk.
11-101-1128 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9525 Supplier MilliporeSigma™ Supelco™ Supplier No. 22475
Add to Cart
Add to Cart
Characteristics: High reusability.Material Composition: DuPont VESPEL SP-1 (100% polyimide)Maximum Temperature: 350°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/4 in.
Diameter (English) Inner 1/4 in.
Packaging Quantity 50/Pk.
Description Ferrule Type: Packed Column
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC