Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2 Packed Column Ferrule, 1/8 in.
Catalog No. 111009526 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
50/Pk.
Catalog No. Packaging Quantity
11-100-9526 50/Pk.
1 options
Catalog No. 11-100-9526 Supplier MilliporeSigma™ Supelco™ Supplier No. 22476
Add to Cart
Add to Cart
Characteristics: High reusability.Material Composition: DuPont VESPEL SP-1 (100% polyimide)Maximum Temperature: 350°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/8 in.
Diameter (English) Inner 1/8 in.
Packaging Quantity 50/Pk.
Description Ferrule Type: Packed Column
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC