Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2A Capillary Ferrule, General Purpose, 0.10-0.25 mm Column I.D. (2-hole)
Catalog No. 111011308 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
5/Pk.
Catalog No. Packaging Quantity
11-101-1308 5/Pk.
1 options
Catalog No. 11-101-1308 Supplier MilliporeSigma™ Supelco™ Supplier No. 22467
Add to Cart
Add to Cart
Characteristics: Seals at 1/4 turn past fingertight. High reusability. Won't stick to metal or glass.Material Composition: DuPont VESPEL SP-21 (85% polyimide, 15% graphite). Maximum Temperature: 400°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.4 mm
Packaging Quantity 5/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC