Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2A Capillary Ferrule, General Purpose, 0.10-0.25 mm Column I.D.
Catalog No. 111009299 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
50/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
11-100-9299 50/Pk.
11-101-0967 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-9299 Supplier MilliporeSigma™ Supelco™ Supplier No. 22474
Add to Cart
Add to Cart
Characteristics: Seals at 1/4 turn past fingertight. High reusability. Won't stick to metal or glass.Material Composition: DuPont VESPEL SP-21 (85% polyimide, 15% graphite). Maximum Temperature: 400°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.4 mm
Packaging Quantity 50/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC