Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2A Packed Column Ferrule, 1/8 in.
Catalog No. 111009297 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
50/Pk.
Catalog No. Packaging Quantity
11-100-9297 50/Pk.
1 options
Catalog No. 11-100-9297 Supplier MilliporeSigma™ Supelco™ Supplier No. 22472
Add to Cart
Add to Cart
Characteristics: Seals at 1/4 turn past fingertight. High reusability. Won't stick to metal or glass. Material Composition: DuPont VESPEL SP-21 (85% polyimide, 15% graphite). Maximum Temperature: 400°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/8 in.
Diameter (English) Inner 1/8 in.
Packaging Quantity 50/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC