Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2A Reducing Ferrule, Column in 1/4 in. fitting
Catalog No. 111010912 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
11-101-0912 10/Pk.
1 options
Catalog No. 11-101-0912 Supplier MilliporeSigma™ Supelco™ Supplier No. 22486
Add to Cart
Add to Cart
Characteristics: Seals at 1/4 turn past fingertight. High reusability. Won't stick to metal or glass. Material Composition: DuPont VESPEL SP-21 (85% polyimide, 15% graphite). Maximum Temperature: 400°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/16 in.
Diameter (English) Inner 1/16 x 1/4 in.
Packaging Quantity 10/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC