Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-2B Double-Tapered Ferrule, 0.10-0.25 mm to like I.D. Column
Catalog No. 111011279 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
2/Pk.
Catalog No. Packaging Quantity
11-101-1279 2/Pk.
1 options
Catalog No. 11-101-1279 Supplier MilliporeSigma™ Supelco™ Supplier No. 22453
Add to Cart
Add to Cart
Max. Temp.: 350°C. Composition: DuPont VESPEL SP-211 (75% polyimide, 15% graphite, 10% PTFE). Characteristics: Conforms easily to capillary column, ensuring an effective seal and less chance of breakage.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 0.4 mm
For Use With (Application) Gas Chromatography (GC)
Packaging Quantity 2/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC