Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-4 Packed Column Ferrule, 1/16 in.
Catalog No. 111011133 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
10/Pk.
Catalog No. Packaging Quantity
11-101-1133 10/Pk.
1 options
Catalog No. 11-101-1133 Supplier MilliporeSigma™ Supelco™ Supplier No. 22495U
Add to Cart
Add to Cart
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation .Material Composition: flexible graphite. Maximum Temperature: 450°C
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Outer 1/16 in.
Diameter (English) Inner 1/16 in.
Packaging Quantity 10/Pk.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC