Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Super Challenger™
SDP

Catalog No. 14727032
Encompass
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-727-032 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

When using the Super Challenger Ball with non-human primates, we recommend adjusting the chain to a shorter length.

Catalog No. 14-727-032 Supplier Bio-Serv™ Supplier No. K3100S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

When using the Super Challenger Ball with non-human primates, we recommend adjusting the chain to a shorter length.

  • Made of sturdy polyethylene and stainless steel
  • Internal disk provides additional challenge
  • Hole size accommodates larger treats
  • Hang from outside of cage for easy filling
  • Cagewasher safe
  • 6.5 in. diameter ball with a 10 in. adjustable stainless steel chain

Specifications

Product Type Primate Enrichment Device
Color Yellow
Material Polyethylene
Length (English) 10 in. chain
Diameter (English) Inside 6.5 in. Ball
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.