Learn More
MilliporeSigma™ Supelco™ Super Clean™ (Base-Plate Design) Gas Purifiers
For the Purification and Management of GC Gas Streams
Supplier:  MilliporeSigma™ Supelco™ SU861027
 View more versions of this product
Catalog No. 11-100-9287
Only null left
Add to Cart
Description
Description
  • Provides gas at the proper purity level and pressure, based on its intended use
  • The purifiers connect into a base plate in such a manner that gas supply is not interrupted during purifier change-out
  • Purifier can be replaced without shutting down the entire system
  • Eliminates the risk of forcing a slug of air into the instrument after purifier change-out
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Enhancing The Performance of GC Detectors
1 Ea.
Triple Trap (Hydrocarbon, Moisture, Oxygen)
GC Detectors
For Helium Carrier Gas
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.