Learn More
MilliporeSigma™ Supelco™ Super Clean™ (Base-Plate Design) Kits
A unique, point-of-use diffusion proof purification system to purify carrier, fuel, and other gases for GC or GC-MS systems
Supplier: MilliporeSigma™ Supelco™ SU861040
Description
- Purifiers remove hydrocarbons, oxygen (color indicated), and moisture (color indicated) to better than 6.0 gas (99.9999%) quality at 2 L/min., independent of the original gas quality
Specifications
Purification of Carrier, Fuel, and Other Gases | |
GC or GC-MS Systems | |
SU861027, and SU861011 | |
Helium Carrier Gas Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Super Clean is a trademark of Scientific Glass Technology B.V.
Spot an opportunity for improvement?Share a Content Correction