Learn More
Thermo Scientific™ Super Clean™ Gas Cartridge Filter Baseplates and Accessories
Catalog No. 60180818
Change view
Click to view available options
Type:
Dual Base
Flush Cap
Four-Position Base
O-Rings for Baseplates
Triple Base
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
60-180-818 Four-Position Base
60-180-815 Dual Base
60180817 Triple Base
60180837 O-Rings for Baseplates
60180838 Flush Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-180-818 Supplier Thermo Scientific™ Supplier No. 60180818
Add to Cart
Add to Cart
Select from a range of quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters.
Use quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters. A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change. The baseplates have a one-time installation procedure. Super Clean Cartridge Filters are not included.
Additional Product Information
  • Use with Super Clean cartridge filters
  • Filters not included
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Moisture, Oxygen, 2x Hydrocarbon
Description For Gas: High Capacity Carrier, Fuel, and Make Up Gas
Type Four-Position Base
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.