Learn More
StatLab SUPER PAP PEN
SDP
Supplier:  StatLab WISPP
Encompass_Preferred
Designed to prevent expensive antibodies from running of slide by forming a hydrophobic barrier around the circled specimen.
Catalog No. NC9993290
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.