Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Super Saucer on Chain
SDP

Catalog No. 14727033
Encompass
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-727-033 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-727-033 Supplier Bio-Serv™ Supplier No. K3607
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Certified (Contaminant Screened) Super Saucer with attached welded link, stainless steel chain with quick links (overall chain length - approximately 6 in.)
  • A multi-use enrichment toy that promotes a variety of natural behaviors
  • Can be used as a visual barrier for submissive monkeys
  • Cagewasher safe
  • 9 in. diameter
  • Overall chain length 6 in.

Specifications

Product Type Enrichment Device
Color Blue
Length (English) 6 in. Chain
Diameter (English) Inside 9 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.