Learn More
Description
- Multi-directional surface cleats effectively remove and trap dirt and sand below shoe level to prevent tracking
- Drainable borders filter water off mat
- Certified high-traction by the National Floor Safety Institute (NFSI)
- Suitable for Harsh Environments
- Grease/oil proof; chemical resistant
- Easy to Clean - Lightweight flexible design ensures easy handling and cleaning (shake off soil and hose off)
- Can be commercially laundered or autoclave sterilized
- 3/16 in. thick nitrile rubber construction; available with holes for additional drainage
Specifications
Specifications
| Width (English) | 5 ft. |
| Quantity | 1 Ea. |
| Dimensions (L x W) | 3 x 5 ft. |
| Certifications/Compliance | NFSI Certified. Made in USA |
| Color | Black |
| Thickness (English) | 3/16 in. |
| Length (English) | 3 ft. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.