Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Super Spring with Stand
SDPSCI_ED

Catalog No. S27851 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Stand
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27851 Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27851 Supplier United Scientific Supplies Supplier No. SSPRING
Add to Cart
Add to Cart

Extra long spring allows for magnification of wave demonstrations

  • Extra-long spring ideal for wave demonstrations
  • Stand prevents tangling or damage during storage
  • Compressed length: 21 cm
  • Activity guide included

Specifications

Product Type Super Spring Wave Demonstrator
For Use With (Application) Magnification of Wave Demonstrations
Includes Stand
Height (Metric) 21 cm (Compressed)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.