Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperBoost™ EverRed and EverBlue Colorimetric HRP Kits

Catalog No. E40966
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 Slides
5 x 150 Slides
Product Type:
EverBlue HRP Kit, Streptavidin
EverBlue HRP Kit, goat anti-mouse IgG
EverBlue HRP Kit, goat anti-rabbit IgG
EverBlue HRP Substrate Kit
EverRed HRP Kit, Streptavidin
EverRed HRP Kit, goat anti-mouse IgG
EverRed HRP Kit, goat anti-rabbit IgG
EverRed HRP Substrate Kit
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
E40966 150 Slides EverBlue HRP Kit, goat anti-mouse IgG
E40964 150 Slides EverBlue HRP Substrate Kit
E40972 5 x 150 Slides EverBlue HRP Substrate Kit
E40968 150 Slides EverBlue HRP Kit, goat anti-rabbit IgG
E40970 150 Slides EverBlue HRP Kit, Streptavidin
E40963 150 Slides EverRed HRP Substrate Kit
E40971 5 x 150 Slides EverRed HRP Substrate Kit
E40965 150 Slides EverRed HRP Kit, goat anti-mouse IgG
E40967 150 Slides EverRed HRP Kit, goat anti-rabbit IgG
E40969 150 Slides EverRed HRP Kit, Streptavidin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. E40966 Supplier Invitrogen™ Supplier No. E40966
Add to Cart
Add to Cart

SuperBoost™ EverRed and EverBlue Colorimetric HRP Kits use poly-HRP-mediated tyramide signal deposition for permanent labeling of a target protein in IHC, enabling sensitive and specific colorimetric tissue staining that is wash resistant, water/organic solvent mountable, and archivable.

SuperBoost™ colorimetric signal anchoring is a highly sensitive, permanent method for the detection of targets in multiplexable colorimetric immunohistochemistry (IHC) experiments. SuperBoost™ technology combines the permanent staining of poly-HRP-mediated tyramide signal deposition with the bright colors of SuperBoost EverRed and EverBlue to discern signal from noise, yielding precision and sensitivity in colorimetric tissue staining applications.

Tyramide signal deposition in the Tyramide SuperBoost™ kits uses the catalytic activity of horseradish peroxidase (HRP) for high-density labeling of a target protein, enabling permanent colorimetric tissue staining with SuperBoost EverRed and/or EverBlue that is wash resistant, water/organic solvent mountable, and archivable. Unlike many other colorimetric IHC staining methods, SuperBoost EverRed and EverBlue staining is covalently attached to the target resulting in a sharp and intense chromogenic signal that is maintained without needing to air-dry tissue or use unconventional dehydration/clearing protocols.

The kits are simple to use and easily adapted to standard IHC experimental protocols using any tissue type, including formalin-fixed, paraffin-embedded (FFPE) and cryopreserved tissue. Tissues labeled using the SuperBoost EverRed and EverBlue kits can be imaged using any type of brightfield microscope, producing high-resolution images.

Features of the SuperBoost EverRed and EverBlue HRP Kits include:
• Permanent colorimetric staining that is also fluorescent, allowing confirmation of the staining pattern with both brightfield and fluorescence imaging
• Compatibility with other common colorimetric counterstains
• Multiplexability between EverRed, EverBlue, and DAB

How does SuperBoost color signal anchoring work?
SuperBoost EverRed and EverBlue HRP Kits use a simple workflow similar to standard IHC staining methods. For SuperBoost EverRed and EverBlue HRP tissue staining, an antigen is detected by a primary antibody, followed by staining with a secondary antibody or streptavidin (for biotinylated primary antibodies) conjugated with poly-HRP. HRP then enzymatically activates a chromogen tyramide, resulting in localized covalent deposition of the activated tyramide derivative at tyrosine residues on and surrounding the protein epitope targeted by the primary antibody. Treatment of the labeled tissue with the EverRed or EverBlue reagent then results in a permanent colorimetric signal at the site of the covalent tyramide deposition.

SuperBoost EverRed/EverBlue HRP Kits, goat anti-mouse IgG/goat anti-rabbit
The SuperBoost EverRed/EverBlue HRP Kits, goat anti-mouse IgG/goat anti-rabbit can be used for permanent red or blue colorimetric detection of mouse or rabbit IgG primary antibodies in IHC applications. The kits include the goat anti-mouse IgG or goat anti-rabbit IgG poly horseradish peroxidase (HRP) conjugate, tyramide HRP substrate, and SuperBoost EverRed or EverBlue reagent along with the buffers and reagents needed for tissue staining and color development. Sufficient reagents are provided to stain 150 tissue sections.

The four options for these kits include:
• SuperBoost™ EverRed HRP Kit, goat anti-mouse IgG
• SuperBoost™ EverRed HRP Kit, goat anti-rabbit IgG
• SuperBoost™ EverBlue HRP Kit, goat anti-mouse IgG
• SuperBoost™ EverBlue HRP Kit, goat anti-rabbit IgG

SuperBoost EverRed/EverBlue HRP Kits, Streptavidin
The SuperBoost EverRed/EverBlue HRP Kits, Streptavidin can be used for permanent red or blue colorimetric detection of biotinylated primary antibodies in IHC applications. The kits include the streptavidin HRP conjugate, tyramide HRP substrate, and SuperBoost EverRed or EverBlue reagent along with the buffers and reagents needed for tissue staining and color development. Sufficient reagents are provided to stain 150 tissue sections.

SuperBoost EverRed/EverBlue HRP Substrate Kits
The SuperBoost EverRed/EverBlue HRP Substrate Kits include the tyramide HRP substrate and SuperBoost EverRed or EverBlue reagent for permanent red or blue colorimetric detection in IHC applications using your own HRP conjugate. Sufficient reagents are provided to stain 150 tissue sections or 5 x 150 tissue sections.

TRUSTED_SUSTAINABILITY

Specifications

Product Type EverBlue HRP Kit, goat anti-mouse IgG
Clonality None
Target Species Mouse
Quantity 150 Slides
Product Line SuperBoost
Content And Storage • 28.5 mL 1X Endogenous peroxide quencher (stabilized 3% H2O2 solution, store at 2–8°C)
• 22.5 mL 1X Blocking buffer (10% goat serum) (2–8°C)
• 22.5 mL 1X Goat anti-mouse secondary antibody, poly HRP conjugate (2–8°C, do not freeze)
• 3 mL 10X Tyramide HRP substrate (2–8°C)
• 20 mL 2X Hydrogen peroxide for reaction buffer (2–8°C, protect from light)
• 25 mL 2X HRP reaction buffer (2–8°C)
• 3 x 2 mg SuperBoost™ EverBlue reagent (2–8°C)
• 30 mL SuperBoost™ EverBlue development buffer (2–8°C)

Shipping Condition Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.