Learn More
Invitrogen™ SuperBoost™ EverRed and EverBlue Colorimetric HRP Kits
Description
SuperBoost™ colorimetric signal anchoring is a highly sensitive, permanent method for the detection of targets in multiplexable colorimetric immunohistochemistry (IHC) experiments. SuperBoost™ technology combines the permanent staining of poly-HRP-mediated tyramide signal deposition with the bright colors of SuperBoost EverRed and EverBlue to discern signal from noise, yielding precision and sensitivity in colorimetric tissue staining applications.
Tyramide signal deposition in the Tyramide SuperBoost™ kits uses the catalytic activity of horseradish peroxidase (HRP) for high-density labeling of a target protein, enabling permanent colorimetric tissue staining with SuperBoost EverRed and/or EverBlue that is wash resistant, water/organic solvent mountable, and archivable. Unlike many other colorimetric IHC staining methods, SuperBoost EverRed and EverBlue staining is covalently attached to the target resulting in a sharp and intense chromogenic signal that is maintained without needing to air-dry tissue or use unconventional dehydration/clearing protocols.
The kits are simple to use and easily adapted to standard IHC experimental protocols using any tissue type, including formalin-fixed, paraffin-embedded (FFPE) and cryopreserved tissue. Tissues labeled using the SuperBoost EverRed and EverBlue kits can be imaged using any type of brightfield microscope, producing high-resolution images.
Features of the SuperBoost EverRed and EverBlue HRP Kits include:
• Permanent colorimetric staining that is also fluorescent, allowing confirmation of the staining pattern with both brightfield and fluorescence imaging
• Compatibility with other common colorimetric counterstains
• Multiplexability between EverRed, EverBlue, and DAB
How does SuperBoost color signal anchoring work?
SuperBoost EverRed and EverBlue HRP Kits use a simple workflow similar to standard IHC staining methods. For SuperBoost EverRed and EverBlue HRP tissue staining, an antigen is detected by a primary antibody, followed by staining with a secondary antibody or streptavidin (for biotinylated primary antibodies) conjugated with poly-HRP. HRP then enzymatically activates a chromogen tyramide, resulting in localized covalent deposition of the activated tyramide derivative at tyrosine residues on and surrounding the protein epitope targeted by the primary antibody. Treatment of the labeled tissue with the EverRed or EverBlue reagent then results in a permanent colorimetric signal at the site of the covalent tyramide deposition.
SuperBoost EverRed/EverBlue HRP Kits, goat anti-mouse IgG/goat anti-rabbit
The SuperBoost EverRed/EverBlue HRP Kits, goat anti-mouse IgG/goat anti-rabbit can be used for permanent red or blue colorimetric detection of mouse or rabbit IgG primary antibodies in IHC applications. The kits include the goat anti-mouse IgG or goat anti-rabbit IgG poly horseradish peroxidase (HRP) conjugate, tyramide HRP substrate, and SuperBoost EverRed or EverBlue reagent along with the buffers and reagents needed for tissue staining and color development. Sufficient reagents are provided to stain 150 tissue sections.
The four options for these kits include:
• SuperBoost™ EverRed HRP Kit, goat anti-mouse IgG
• SuperBoost™ EverRed HRP Kit, goat anti-rabbit IgG
• SuperBoost™ EverBlue HRP Kit, goat anti-mouse IgG
• SuperBoost™ EverBlue HRP Kit, goat anti-rabbit IgG
SuperBoost EverRed/EverBlue HRP Kits, Streptavidin
The SuperBoost EverRed/EverBlue HRP Kits, Streptavidin can be used for permanent red or blue colorimetric detection of biotinylated primary antibodies in IHC applications. The kits include the streptavidin HRP conjugate, tyramide HRP substrate, and SuperBoost EverRed or EverBlue reagent along with the buffers and reagents needed for tissue staining and color development. Sufficient reagents are provided to stain 150 tissue sections.
SuperBoost EverRed/EverBlue HRP Substrate Kits
The SuperBoost EverRed/EverBlue HRP Substrate Kits include the tyramide HRP substrate and SuperBoost EverRed or EverBlue reagent for permanent red or blue colorimetric detection in IHC applications using your own HRP conjugate. Sufficient reagents are provided to stain 150 tissue sections or 5 x 150 tissue sections.
Specifications
Specifications
| Product Type | EverBlue HRP Kit, goat anti-rabbit IgG |
| Clonality | None |
| Target Species | Rabbit |
| Quantity | 150 Slides |
| Product Line | SuperBoost |
| Content And Storage | • 28.5 mL 1X Endogenous peroxide quencher (stabilized 3% H2O2 solution, store at 2–8°C) • 22.5 mL 1X Blocking buffer (10% goat serum) (2–8°C) • 22.5 mL 1X Goat anti-rabbit secondary antibody, poly HRP conjugate (2–8°C, do not freeze) • 3 mL 10X Tyramide HRP substrate (2–8°C) • 20 mL 2X Hydrogen peroxide for reaction buffer (2–8°C, protect from light) • 25 mL 2X HRP reaction buffer (2–8°C) • 3 x 2 mg SuperBoost™ EverBlue reagent (2–8°C) • 30 mL SuperBoost™ EverBlue development buffer (2–8°C) |
| Shipping Condition | Wet Ice |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.