Learn More
TOPOGEN INC Supercoiled pHOT-1 DNA Plasmid
Supplier:  TOPOGEN INC TG20303
Encompass_Preferred
This plasmid is ideal for assaying any topoisomerase. pHOT-1 is relatively small which is ideal for displaying intermediate topoisomers. Since all topoisomerases display degenerate DNA binding and cleavage sequences, pHOT-1 will support activity for a wide array of topoisomerase. Supercoiled pHOT-1 is a scaled-down derivative of pBR322 and optimized for higher yields. It has one additional feature for researchers who wish to assess human topo I cleavage at a single sequence: we have included the human topoisomerase I preferential binding site (the hexadecameric sequence described by Westergaard and co-workers, see Bonven et al., 1985) that is derived from the tetrahymena ribosomal gene repeat. This construct is useful as a topo I cleavage substrate since the enzyme efficiently cleaves at the hexadecameric site in the absence of camptothecin. This allows the investigator to rigorously evaluate new potential topo I inhibitors because a single background cleavage is a built-in control.
Catalog No. NC2216101
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.