Learn More
Cytiva Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns
GSA_VA
Catalog No. 45002570 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume:
25 to 500μL
Catalog No. Bed Volume
45-002-570 25 to 500μL
1 options
Catalog No. 45-002-570 Supplier Cytiva Supplier No. 28990944
Add to Cart
Add to Cart
For size exclusion chromatography and analysis of proteins with molecular weights between 10,000 and 600,000. Cytiva Lifescience™ Superdex™ 200 Increase 10/300 GL Prepacked Tricorn™ Columns are designed for small scale preparative purification.
  • For small-scale (25 to 500μL) preparative purification and for analytical purposes, suited for antibodies
  • Allows characterization of proteins, aggregate analysis of antibodies, size homogeneity analysis of membrane proteins, or protein-protein interactions
  • Prepacked with Superdex 200 Increase media
  • Composite of cross-linked high-flow agarose and dextran base matrix for high flow rates
  • Smaller bead size of average 8.6μm for increased resolution
  • Flow Rate: <1.80mL/min1
  • Bed Volume: Approximately 24mL
  • Column 1.D.: 10mm
  • Columns are supplied with two fingertight connectors 1/16 in. male for connection to AKTA™ or other system
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Bed Volume 25 to 500μL
Product Type Superdex Pre-packed Column
For Use With (Application) Small-scale (25 to 500μL) preparative purification and analysis
Material Cross-linked agarose and dextran