Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Superdex™ 200 Prep Grade
GSA_VA

Catalog No. 45002039 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
150mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-039 150mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-039 Supplier Cytiva Supplier No. 17104301
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Produce separation medium with high selectivity and resolution. Cytiva Life Sciences™ Superdex™ 200 Prep Grade for purifying proteins of fractionation range: 10000 to 600000D

  • Gel filtration medium
  • For purifying proteins of fractionation range: 10000 to 600000Da
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
  • pH stability working range
  • Available in 150mL bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 150mL
Flow Rate 10 to 50 cm/hr.
Particle Size 24 to 44 μm
Product Type Superdex Media
Format Solid Spherical Bead
For Use With (Application) Separation, polishing and product formulation of Monoclonal Antibodies
Max. Pressure 70psi
pH Range 3 to 13
Support Type Composite of cross-linked Agarose and Dextran
Sample Type Recombinant Protein
Content And Storage 4 to 30°C, 20% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.