Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Superdex 30 PG size exclusion chromatography media prepacked into HiLoad 26/600 column
GSA_VA

Catalog No. 45002487 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume:
320 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
45-002-487 320 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-487 Supplier Cytiva Supplier No. 28989332
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Includes

Dismantling tool, support screen, net ring (10μm), O-ring, stop plugs, and instructions.

Superdex 30 prep grade (PG) size exclusion chromatography (gel filtration) media prepacked into a HiLoad 26/600 column is optimized for the separation of proteins and peptides, but can also be used with success to separate oligosaccharides.

  • High-resolution separation – with short run times and good recovery.
  • High capacity – at high flow rates.
  • Prepacked – for convenient ready-to-go use and reproducible results.
  • Scalable protein separation – easily scale-up separations to production levels.
  • Integrated workflows – 1/16 in. fittings for convenient connection with ÄKTA protein purification systems.

Recommended Storage

4 to 30°C, 20% Ethanol

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Flow Rate < 2.6 mL/min.
Particle Size 34 μm
Bed Volume 320 mL
Product Type Superdex Pre-packed Column
Diameter (Metric) Inner 26 mm
Material Superdex 30 prep grade
Max. Pressure 3 bar [0.3 MPa] (42 psi)
pH Range 3 to 12
Includes Dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs, and instructions.
Packaging 1 × 320mL
Content And Storage 4 to 30°C, 20% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.