Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SuperFi™ Buffers

Catalog No. p-200085140
Encompass_Preferred
Change view
Click to view available options
Color:
Colorless
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12-355-005 Colorless
12-356-005 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-355-005 Supplier Invitrogen™ Supplier No. 12355005
Only null left
Add to Cart
Add to Cart

Invitrogen SuperFi Buffer is a 5X buffer solution optimized for use with Platinum SuperFi DNA Polymerase.

Invitrogen™ SuperFi™ Buffer is a 5X buffer solution optimized for use with Invitrogen™ Platinum™ SuperFi DNA Polymerase. The buffer contains 7.5 mM MgCl2 which provides 1.5 mM MgCl2 in the final reaction.

The SuperFi Green Buffer (Cat. No. 12356005) includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with the enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless SuperFi™ Buffer. (12355005)

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
For Use With (Application) Hot-start PCR
Quantity 4 x 1.25 mL
Includes 5X SuperFi Buffer (4 x 1.25 mL)

Store at –5°C to –30°C.
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.