Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific SuperFlex™ 'Break-Away' Cap Strips for 96-well PCR Plates

Catalog No. 23045583 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-583 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-045-583 Supplier Simport™ Scientific Supplier No. T32196N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Break-Away' Cap Strips provide closure for your PCR plates in convenient, 8-well strips.

  • For use with natural or white PCR plates
  • Strips of 8 closure caps
  • Securely seals wells

Specifications

Barcode No Barcode
Array 8 x 12
Autoclavable Autoclavable
Color Clear
Disposable Single-use
Product Type Closure Strip
For Use With (Equipment) SuperFlex™ 'Break-Away' 96-well PCR Plates
Material Polypropylene
Sterility Non-sterile
For Use With (Application) PCR and qPCR
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.