Learn More
Research Products International Corp Superflex Break Awy Plate
SDP
Supplier:  Research Products International Corp 400701S
Encompass_Preferred
Eliminate the use of scissors that cause perforated wells, damage sealing rings, and could risk contamination issues. The wells (0.2ml) can be separated into 8 strips and are more economical than individual strips. The wells are individually flexible to have a perfect fit and compatible with all instruments that fit non-skirted, low profile plates. SuperFlex White “Break-Away” PCR Plate improves signal reflection, reduces variation and increases the sensitivity of qPCR assays. Sterile plates are gamma irradiated.
Catalog No. 50-238-8392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.