Learn More
Simport™ Scientific SUPERFLO™ Histology Micro Biopsy Cassette, Hinged Design, Sleeve
Catalog No. 23045660 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-660 Aqua
23-045-650 White
23-045-651 Pink
23-045-652 Green
23-045-653 Yellow
23-045-654 Blue
23-045-655 Peach
23-045-656 Tan
23-045-657 Gray
23-045-658 Lilac
23-045-659 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 23-045-660 Supplier Simport™ Scientific Supplier No. M53512SL
Add to Cart
Add to Cart
This item is not returnable. View return policy
This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface. Molded from high-density polymer, keeps specimens safely submerged in liquid.
  • Unique lateral vents, fluids can access tissue from all 4 sides
  • Basket style for small tissue management
  • Special hinged cover design with front opening
  • Smooth 45° angle writing surface
  • Available in 11 colors
  • This cassette model is supplied in plastic sleeves suitable for printers
  • IVDR
  • CE
Specifications
Pore Style 0.30 x 0.30 mm
Quantity 10 Sleeves of 75
Packaging Type Sleeve
Color Aqua
Product Type Micro Biopsy Cassette