Learn More
Simport™ Scientific SUPERFLO™ Histology Micro Biopsy Cassette, Hinged Design, Taped
Catalog No. 23045627 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-627 Aqua
23-045-617 White
23-045-618 Pink
23-045-619 Green
23-045-620 Yellow
23-045-621 Blue
23-045-622 Peach
23-045-623 Tan
23-045-624 Gray
23-045-625 Lilac
23-045-626 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 23-045-627 Supplier Simport™ Scientific Supplier No. M43512T
Add to Cart
Add to Cart
This item is not returnable. View return policy
This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface. Molded from high-density polymer, keeps specimens safely submerged in liquid.
  • Unique lateral vents, fluids can access tissue from all 4 sides
  • Basket style for small tissue management
  • Special hinged cover design with front opening
  • Smooth 45° angle writing surface
  • Available in 11 colors
  • This cassette model is supplied in taped stacks for use with ink jet printers
  • IVDR
  • CE
Specifications
Pore Style 0.64 x 0.64 mm
Packaging Type Taped
Quantity 50 Stacks of 40
Color Aqua
Product Type Micro Biopsy Cassette