Learn More
Simport™ Scientific SUPERFLO™ Histology Tissue Cassette, Hinged Design, Sleeve
Catalog No. 23045682 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-682 Aqua
23-045-672 White
23-045-673 Pink
23-045-674 Green
23-045-675 Yellow
23-045-676 Blue
23-045-677 Peach
23-045-678 Tan
23-045-679 Gray
23-045-680 Lilac
23-045-681 Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 23-045-682 Supplier Simport™ Scientific Supplier No. M53312SL
Add to Cart
Add to Cart
This item is not returnable. View return policy
This model has a special hinge that holds the base and the cover together. The hinge design allows the cassette to be opened and closed as often as required. It is a front-opening cassette with a 45° smooth writing surface. Molded from high-density polymer, keeps specimens safely submerged in liquid.
  • Unique lateral vents, fluids can access tissue from all 4 sides
  • Basket style for small tissue management
  • Special hinged cover design with front opening
  • Smooth 45° angle writing surface
  • Available in 11 colors
  • This cassette model is supplied in plastic sleeves suitable for printers
  • IVDR
  • CE
Specifications
Pore Style 0.91 x 3.67 mm
Quantity 10 Sleeves of 75
Packaging Type Sleeve
Color Aqua
Product Type Tissue Cassette