Learn More
NOTRAX™ Superfoam™ Anti-Fatigue Mat
Superfoam™ Anti-Fatigue Foam Mat is extremely lightweight and durable and designed especially for ease of handling and ergonomic comfort
Supplier:  NOTRAX™ T17S0032BL
 View more versions of this product
Catalog No. 19-086-229
Only null left
Add to Cart
Description
Description
  • Highly resistant to grease, oils and animal and vegetable fats
  • Closed cell nitrile foam, does not absorb liquids
  • Durable and abrasion resistant
  • Solid for dry areas or perforated drainage holes to remove liquid and debris for worker platform
  • PVC/Nitrile closed cell foam blend
  • Non-allergenic, silicone-free compound
  • 45° Beveled edges on all four sides
Specifications
Specifications
3 x 2 ft.
1 Ea.
3 ft.
5/8 in.
2 ft.
Solid Anti-fatigue Floor Mat
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.