Description
- Made from a completely closed cell foam blend
- Does not absorb liquids
- Highly resistant to grease, oils and animal and vegetable fats
- Durable, light weight rubber is easy to pick up and move
- Ergonomically designed to provide high level of rebound without bottoming out
- Non-allergenic, silicone-free compound
- Beveled edges on all 4 sided
Specifications
Specifications
| Color | Black |
| Length (English) | 36 in. |
| Width (English) | 40 in. |
| Quantity | 1 Ea. |