Learn More
Superior Glove™ Clutch Gear™ Impact-Resistant Mechanics Gloves with PVC Sure Grip™
Description
- Impact-resistant back-of-hand protection
- Protects against knocks and bumps without sacrificing flexibility or dexterity
- PVC Sure Grip™ palm patches enhance grip and improve fingertip protection
- Reinforced thumb patch prevents degradation in high-stress areas
- Adjustable cuffs for additional protection and comfort
Specifications
Specifications
| Coating Material | PVC |
| Color | Yellow, Black |
| Cut Resistance Rating | ANSI A2 |
| Hand | Specific |
| Product Type | Mechanic's Gloves |
| Size | Large |
| For Use With (Application) | Mechanics, Construction, Oil/Gas Drilling and Material Handling |
| Puncture Rating | ASTM F2878 LEVEL 3 |
| Series | MXVSB |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.