Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Clutch Gear™ Impact-Resistant Mechanics Gloves with PVC Sure Grip™

Catalog No. 17400126
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-400-126 X-Large
17-400-123 Small
17-400-124 Medium
17-400-125 Large
17-400-127 2X-Large
17-400-128 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-400-126 Supplier Superior Glove™ Supplier No. MXVSB/XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

These gloves fully protect the back of the hand from impacts, without sacrificing dexterity or flexibility. PVC Sure Grip™ palm patches enhance grip capabilities.

  • Impact-resistant back-of-hand protection
  • Protects against knocks and bumps without sacrificing flexibility or dexterity
  • PVC Sure Grip™ palm patches enhance grip and improve fingertip protection
  • Reinforced thumb patch prevents degradation in high-stress areas
  • Adjustable cuffs for additional protection and comfort

Specifications

Coating Material PVC
Color Yellow, Black
Cut Resistance Rating ANSI A2
Hand Specific
Product Type Mechanic's Gloves
Size X-Large
For Use With (Application) Mechanics, Construction, Oil/Gas Drilling and Material Handling
Puncture Rating ASTM F2878 LEVEL 3
Series MXVSB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.