Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Contender™ Terry-Knit Metal-Stamping Gloves

Catalog No. 17401262
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-262 Medium
17-401-263 Large
17-401-264 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-401-262 Supplier Superior Glove™ Supplier No. TRFGK/M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Terry-knit aramid fiber gloves provide abrasion and cut protection. Ideal especially for metal stamping applications, but can also be used in automotive, assembly and material handling industries.

  • Terry knit contains composite filament fiber for abrasion and cut protection
  • Terry loops enhance cut resistance
  • Longer knit-wrist cuffs are fitting for stay-in-place comfort
  • Cotton blend absorbs surface oil/grease to increase grip on metal surfaces

Specifications

ANSI Abrasion Rating ANSI 2
Certifications/Compliance ANSI 5 Heat
Construction Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Stainless Steel Fiber, Nylon
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) heavy Metal stamping, automotive press and welding, automotive, parts assembly, material handling
Glove Cuff Style Knit
Model TRFGK/M
Series Contender™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.