Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Endura™ Premium Cut-Resistant Fitter Gloves

Catalog No. 17401274
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-274 X-Large
17-401-271 Small
17-401-272 Medium
17-401-273 Large
17-401-275 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-401-274 Supplier Superior Glove™ Supplier No. 69BSKFFL/X
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cut-resistant fitter gloves feature a para-aramid liner for both ANSI Level A5 cut and puncture protection.

  • Blended para-aramid liner for 360° cut protection
  • Heavy-duty premium split-leather is ideal for tough jobs
  • ANSI level 5 puncture resistance
  • Sewn with strong, heat-resistant para-aramid thread for seam integrity
  • Gunn-cut pattern moves seams away from between fingers and toward the back for less rubbing
  • Backs feature leather knuckle straps and elasticized wrists for a snug fit
  • Palms feature 1.4 mm. premium side-split leather

Specifications

Color Gray, Tan
Cut Resistance Rating ANSI A5
Material Leather
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Metal fabrication, glass handling, maintenance, automotive, construction, stamping, mining, forestry, steel foundries
Liner Material Aramid Fiber
Model 69BSKFFL/XL
Puncture Rating ANSI/ISEA 105-16 Level 5
Series Endura™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.