Learn More
Superior Glove™ Emerald CX™ Aramid Fiber Composite Knit Gloves, Micropore Nitrile Coating
Catalog No. 17400114
Change view
Click to view available options
Size:
6
7
8
9
10
11
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-400-114 7
17-400-113 6
17-400-115 8
17-400-116 9
17-400-117 10
17-400-118 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-400-114 Supplier Superior Glove™ Supplier No. S13CXPNT7
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cut-resistant gloves offer a blend of aramid fiber and stainless steel with micropore nitrile palm coating.
  • 13-gauge blend of aramid fiber and stainless steel
  • Micropore nitrile palm coating does not absorb oils and liquids so hands stay dry
  • Craters act like tiny suction cups to improve grip in wet and oily conditions
  • Nitrile thumb crotch reinforcement to reduce wear in critical areas
  • Touchscreen compatible
Specifications
ANSI Abrasion Rating ANSI 4
Coating Material Microporous Nitrile
Color Gray, Black
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Steel Fiber
Product Type Cut-resistant Gloves
Size 7
For Use With (Application) Handling sharp, Oily Metal parts, circuit boards, automotive, Metal fabrication, assembly, sheet Metal work, Metal stamping, small parts handling, Glass manufacturing, window manufacturing
Glove Cuff Style Knit
Model S13CXPNT/7
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Emerald CX™
Yarn Gauge 13