Learn More
Superior Glove™ Emerald CX™ Aramid Fiber Composite Knit Gloves, Micropore Nitrile Coating
Description
- 13-gauge blend of aramid fiber and stainless steel
- Micropore nitrile palm coating does not absorb oils and liquids so hands stay dry
- Craters act like tiny suction cups to improve grip in wet and oily conditions
- Nitrile thumb crotch reinforcement to reduce wear in critical areas
- Touchscreen compatible
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Coating Material | Microporous Nitrile |
| Color | Gray, Black |
| Cut Resistance Rating | ANSI A4 |
| Material | Aramid Fiber, Steel Fiber |
| Product Type | Cut-resistant Gloves |
| Size | 9 |
| For Use With (Application) | Handling sharp, Oily Metal parts, circuit boards, automotive, Metal fabrication, assembly, sheet Metal work, Metal stamping, small parts handling, Glass manufacturing, window manufacturing |
| Glove Cuff Style | Knit |
| Model | S13CXPNT/9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.