Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Emerald CX™ Aramid Fiber Blend Gloves, Polyurethane Palm Coating

Catalog No. 17401237
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-237 11
17-401-232 6
17-401-233 7
17-401-235 9
17-401-236 10
17-401-238 12
17-401-234 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-401-237 Supplier Superior Glove™ Supplier No. S13CXPU11
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cut-resistant gloves with a knit blend of aramid fiber and stainless steel. Polyurethane palm coating provides dexterity and tactility.

  • Knitted with a blend of aramid fiber and stainless steel for high cut resistance
  • 13-gauge knit provides a balance between high dexterity and longevity
  • Polyurethane palm coating offers dexterity and tactile sensitivity while providing comfort and secure grip
  • Ideal for glass handling, non-marring gloves will not leave residue, smudges or fingerprints
  • Speckled color hides dirt for longer life span

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size 11
For Use With (Application) Sharp steel, circuit boards, automotive, glass industry, metal fabrication, assembly, sheet metal work, HVAC
Glove Cuff Style Knit
Model S13CXPU/11
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Emerald CX™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.