Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Emerald CX™ Cut-Resistant Gloves, Leather Palm

Catalog No. 17401222
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-222 X-Small
17-401-223 Small
17-401-224 Medium
17-401-225 Large
17-401-226 X-Large
17-401-227 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-401-222 Supplier Superior Glove™ Supplier No. SKWCPLP/XS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Aramid fiber/stainless steel string knit gloves with leather palm offer comfort and durability.

  • Aramid fiber/stainless steel composite yarn blended with polyester
  • Comfortable and durable
  • 7-gauge, leather palm
  • Ideal for moderate cut applications such as handling metal stampings, glass or other sharp materials

Specifications

Certifications/Compliance ANSI 3 Heat
Coating Material Leather
Color Yellow, Black, Gray
Cut Resistance Rating ANSI A3
Material Aramid Fiber, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size X-Small
For Use With (Application) Sheet metal, metal stamping and fabrication, glass handling
Glove Cuff Style Knit
Model SKWCPLP/XS
Puncture Rating ANSI/ISEA 105-16 Level 5
Series Emerald CX™
Yarn Gauge 7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.