Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Emerald CX™ Nylon/Stainless Steel Cut-Resistant Gloves, Foam Nitrile Coating

Catalog No. 17401216
Change view
Click to view available options
Size:
6
7
8
9
10
11
12
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-216 7
17-401-215 6
17-401-217 8
17-401-218 9
17-401-219 10
17-401-220 11
17-401-221 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-401-216 Supplier Superior Glove™ Supplier No. S13KBFNT7
This item is not returnable. View return policy

Combination of nylon and fine-gauge steel for cut protection when handling sharp steel or glass.

  • Combination of nylon and fine-gauge steel
  • 13-gauge; cut, abrasion and puncture resistant
  • Extra-long knit wrists protect wrists, keep gloves fitting comfortably
  • Lint-free nylon is lightweight and tough for better dexterity
  • Good touch sensitivity
  • Ideal for handling sharp steel or glass

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A2
Material Nylon, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size 7
For Use With (Application) HVAC, automotive, sharp Steel, Glass, Metal fabrication, light Metal stamping
Glove Cuff Style Knit
Model S13KBFNT/7
Puncture Rating ANSI/ISEA 105-16 Level 3
Series Emerald CX™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.