Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Sure Knit™ Dyneema™ Cut-Resistant Gloves

Catalog No. 17401269
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Green
Purple
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Glove Cuff Color
17-401-269 X-Large Green
17-401-265 X-Small Purple
17-401-267 Medium Yellow
17-401-268 Large White
17-401-270 2X-Large Black
17-401-266 Small Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-401-269 Supplier Superior Glove™ Supplier No. S13GDSTL/X
Add to Cart
Add to Cart
This item is not returnable. View return policy

Blend of Dyneema™ and stainless steel gloves offer ANSI cut level A5 resistance.

  • Blend of Dyneema™ and stainless steel
  • ANSI cut-level A5
  • Fine 13-gauge knit reduces hand fatigue for all-day comfort and touch sensitivity
  • Treated with AlphaSan™, a silver-based antimicrobial agent that prevents and impedes bacterial growth
  • Anti-microbial agent does not impart any taste to food being handled
  • Touchscreen compatible

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Uncoated
Color Gray
Cut Resistance Rating ANSI A4
Material Dyneema, Stainless Steel Fiber, Spandex
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) meat processing, food and beverage, handling sharp materials, solar panel manufacturing
Glove Cuff Color Green
Glove Cuff Style Knit
Hem Color Green
Model S13GDSTL/XL
Series Sure Knit™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.