Learn More
Superior Glove™ TenActiv™ String Knit Cut-Resistant Gloves, Foam Nitrile Coating
Catalog No. 17401259
Change view
Click to view available options
Size:
6
7
9
10
11
12
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-401-259 10
17-401-255 6
17-401-256 7
17-401-258 9
17-401-261 12
17-401-260 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-401-259 Supplier Superior Glove™ Supplier No. STAFGFNT10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Knit with high-tenacity composite yarns designed to meet a range of applications.
  • TenActiv™ yarn is comfortable, breathable and will not absorb sweat/moisture
  • 13-gauge knit offers great balance between high dexterity and high abrasion resistance
  • Foam nitrile palm coating provides good grip for lightly oiled parts
  • Nitrile thumb crotch reinforcement to reduce wear in critical areas
  • Touch screen compatible
  • ASTM ANSI Cut Level 4
Specifications
ANSI Abrasion Rating ANSI 4
Coating Material Nitrile Foam
Color Black
Cut Resistance Rating ANSI A2
Material Composite Fiber
Product Type Cut-resistant Gloves
Size 10
For Use With (Application) Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping
Glove Cuff Style Knit
Model STAFGFNT/10
Puncture Rating ANSI/ISEA 105-16 Level 3
Series TenActiv™
Yarn Gauge 13