Learn More
Superior Glove™ TenActiv™ String Knit Cut-Resistant Gloves, Foam Nitrile Coating
Description
- TenActiv™ yarn is comfortable, breathable and will not absorb sweat/moisture
- 13-gauge knit offers great balance between high dexterity and high abrasion resistance
- Foam nitrile palm coating provides good grip for lightly oiled parts
- Nitrile thumb crotch reinforcement to reduce wear in critical areas
- Touch screen compatible
- ASTM ANSI Cut Level 4
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Coating Material | Nitrile Foam |
| Color | Black |
| Cut Resistance Rating | ANSI A2 |
| Material | Composite Fiber |
| Product Type | Cut-resistant Gloves |
| Size | 11 |
| For Use With (Application) | Handling sharp objects, Glass plants, construction, automotive industry, aerospace industry, Metal fabrication, Metal stamping |
| Glove Cuff Style | Knit |
| Model | STAFGFNT/11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.