Learn More
Cytiva SUPERLOOP 10 ML AKTA
Supplier:  Cytiva 18111381
Encompass
Superloop allows the introduction of large sample volumes into a pressurized fluid system. It is used together with the injection valve MV-7, INV-907, V9-Inj, or V9H-Inj and replaces a simple sample loop.
Catalog No. 50-105-5652