Learn More
Cytiva SUPERLOOP 50 ML AKTA
Supplier:  Cytiva 18111382
Encompass
Superloop allows the introduction of large sample volumes into a pressurized fluid system. It is used together with the injection valve MV-7 or INV-907 and replaces a simple sample loop.
Catalog No. 50-105-5653